2,4,5-Trifluorobenzyl alcohol ≥97%
Supplier: Matrix Scientific
003644-1G
003644-5G
101786-004EA
18.46
USD
101786-004
101786-006
2,4,5-Trifluorobenzyl alcohol ≥97%
2,4,5-Trifluorobenzyl alcohol
Hazard code(s): Irritant
Formula:
C₇H₅F₃O MW: 162.11 g/mol |
MDL Number:
MFCD00061206 CAS Number: 144284-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...