2,3,6-Trifluorobenzyl alcohol ≥97%

Supplier: Matrix Scientific

003643-1G 003643-5G
101786-000EA 581.54 USD
101786-000 101786-002
2,3,6-Trifluorobenzyl alcohol ≥97%
2,3,6-Trifluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₅F₃O
Melting Pt: 43…45 °C
MDL Number: MFCD00061205
CAS Number: 114152-19-1

Order Now

Specification Test Results

Melting point 43-45°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR