2,3,4-Trifluorobenzyl alcohol ≥97%
Supplier: Matrix Scientific
003642-1G
003642-5G
101785-996EA
26.15
USD
101785-996
101785-998
2,3,4-Trifluorobenzyl alcohol ≥97%
2,3,4-Trifluorobenzyl alcohol
Hazard code(s): Irritant
Formula:
C₇H₅F₃O MW: 162.11 g/mol |
MDL Number:
MFCD00083551 CAS Number: 144284-24-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...