α,α,α,3,4,5-Hexafluorotoluene ≥97%

Supplier: Matrix Scientific
Warning

003639-1G 003639-5G
101785-992EA 56.92 USD
101785-992 101785-994
α,α,α,3,4,5-Hexafluorotoluene ≥97%
α,α,α,3,4,5-Hexafluorotoluene

Hazard code(s): Irritant

Formula: C₇H₂F₆
MW: 200.08 g/mol
MDL Number: MFCD00153280
CAS Number: 80172-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR