2,4,6-Trifluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

003636-5G 003636-25G
101785-984EA 23.08 USD
101785-984 101785-986
2,4,6-Trifluorobenzoic acid ≥98%
2,4,6-Trifluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₃F₃O₂
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 143…145 °C
MDL Number: MFCD00042398
CAS Number: 28314-80-9

Order Now

Specification Test Results

Melting point 143-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR