3,4,5-Trifluorobenzamide ≥97%

Supplier: Matrix Scientific

003634-1G 003634-5G
101785-980EA 30.77 USD
101785-980 101785-982
3,4,5-Trifluorobenzamide ≥97%
3,4,5-Trifluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₄F₃NO
MW: 175.11 g/mol
MDL Number: MFCD00083524
CAS Number: 78324-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR