2,4,6-Trifluorobenzamide ≥97%

Supplier: Matrix Scientific

003633-1G 003633-5G
101785-976EA 36.92 USD
101785-976 101785-978
2,4,6-Trifluorobenzamide ≥97%
2,4,6-Trifluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₄F₃NO
MDL Number: MFCD00061199
CAS Number: 82019-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR