2,4,5-Trifluorobenzamide ≥97%

Supplier: Matrix Scientific

003632-1G 003632-5G
101785-972EA 18.46 USD
101785-972 101785-974
2,4,5-Trifluorobenzamide ≥97%
2,4,5-Trifluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₄F₃NO
MDL Number: MFCD00082478
CAS Number: 98349-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR