2,3,5-Trifluorobenzoic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
003626-1G
003626-5G
101785-954EA
69.23
USD
101785-954
101785-956
2,3,5-Trifluorobenzoic acid ≥98%
2,3,5-Trifluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₃F₃O₂ Melting Pt: 108…114 °C |
MDL Number:
MFCD00061200 CAS Number: 654-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...