2,3,4-Trifluorobenzoic acid ≥98% for synthesis
Supplier: Matrix Scientific
|
Warning
|
003625-25G
003625-100G
101785-950EA
95.38
USD
101785-950
101785-952
2,3,4-Trifluorobenzoic acid ≥98% for synthesis
2,3,4-Trifluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₃F₃O₂ Boiling Pt: 237 °C (1013 hPa) Melting Pt: 140 - 142 °C |
MDL Number:
MFCD00061232 CAS Number: 61079-72-9 |
Specification Test Results
Melting point | 140-142°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...