2,4,5-Trifluorobenzoic acid ≥99%

Supplier: Matrix Scientific
Warning

003624-25G 003624-100G
101785-946EA 21.54 USD
101785-946 101785-948
2,4,5-Trifluorobenzoic acid ≥99%
2,4,5-Trifluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₃F₃O₂
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00013306
CAS Number: 446-17-3

Order Now

Specification Test Results

Melting point 99-101°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR