2,4,6-Trifluoroanisole
Supplier: Matrix Scientific
Synonyms:
1,3,5-Trifluoro-2-methoxybenzene, 1-Methoxy-2,4,6-trifluorobenzene
003620-1G
003620-5G
101785-942EA
38.46
USD
101785-942
101785-944
2,4,6-Trifluoroanisole
2,4,6-Trifluoroanisole
Formula:
C₇H₅F₃O MW: 162.11 g/mol |
MDL Number:
MFCD01631502 CAS Number: 219998-30-8 UN: 3271 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...