2,3,4-Trifluoroaniline ≥98%

Supplier: Matrix Scientific
Danger

003616-25G 003616-100G
101785-928EA 21.54 USD
101785-928 101785-930
2,3,4-Trifluoroaniline ≥98%
2,3,4-Trifluoroaniline

Hazard code(s): Irritant

Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: 14 - 15 °C
Density: 1.39 g/cm³ (20 °C)
MDL Number: MFCD00011737
CAS Number: 3862-73-5
EINECS: 407-170-9
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Boiling point 92°C/48mm
Density 1.393
ND20 1.487

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR