3,4,5-Trifluoroaniline ≥98%

Supplier: Matrix Scientific
Warning

003614-5G 003614-25G
101785-922EA 32.31 USD
101785-922 101785-924
3,4,5-Trifluoroaniline ≥98%
3,4,5-Trifluoroaniline

Hazard code(s): Irritant

Formula: C₆H₄F₃N
MW: 147.1 g/mol
MDL Number: MFCD00083522
CAS Number: 163733-96-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR