Trifluoroacetamide ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2,2,2-Trifluoroacetamide

003603-100G 003603-500G
101785-916EA 29.23 USD
101785-916 101785-918
Trifluoroacetamide ≥97%
Trifluoroacetamide

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₂H₂F₃NO
MW: 113.04 g/mol
Boiling Pt: 162.5 °C
Melting Pt: 70…75 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00008008
CAS Number: 354-38-1
EINECS: 206-559-9

Order Now

Specification Test Results

Boiling point 162.5°C
Melting point 70-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR