2,4,5-Trifluoro-3-methoxybenzoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: Moxidectin Impurity 5, 3-Methoxy-2,4,5-trifluorobenzoic acid

003586-25G 003586-100G 003586-250G
101785-890EA 21.54 USD
101785-890 101785-892 101785-894
2,4,5-Trifluoro-3-methoxybenzoic acid ≥98%
2,4,5-Trifluoro-3-methoxybenzoic acid

Hazard code(s): Irritant

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
MDL Number: MFCD00153201
CAS Number: 112811-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR