1H,1H-Heptadecafluoro-1-nonanol ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 1H,1H-Perfluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluoro-1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-Heptadecafluor-1-nonanol

003559-5G 003559-25G
101785-876EA 36.92 USD
101785-876 101785-878
1H,1H-Heptadecafluoro-1-nonanol ≥95% for synthesis
1H,1H-Heptadecafluoro-1-nonanol

Hazard code(s): Irritant

Formula: C₉H₃F₁₇O
Boiling Pt: 84 °C (16mm)
Melting Pt: 68…69 °C
MDL Number: MFCD00153183
CAS Number: 423-56-3

Order Now

Specification Test Results

Boiling point 84°C/16mm
Melting point 68-69°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR