1H,1H-Nonadecafluoro-1-decanol ≥98%

Supplier: Matrix Scientific

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluoro-1-decanol, 1H,1H-Perfluoro-1-decanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluor-1-decanol

003556-5G 003556-25G
101785-866EA 172.31 USD
101785-866 101785-868
1H,1H-Nonadecafluoro-1-decanol ≥98%
1H,1H-Nonadecafluoro-1-decanol

Hazard code(s): Irritant

Formula: C₁₀H₃F₁₉O
MW: 500.1 g/mol
Boiling Pt: 95 °C (20mm)
Melting Pt: 82…84 °C
MDL Number: MFCD00153234
CAS Number: 307-37-9

Order Now

Specification Test Results

Boiling point 95°C/20mm
Melting point 82-84°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR