5-Methylcaproic acid ≥97%

Supplier: Matrix Scientific
Danger

Synonyms: 5-Methylhexanoic acid, 5-Methylhexanoicacid

003527-5G 003527-25G 003527-100G
101785-840EA 41.54 USD
101785-840 101785-842 101785-844
5-Methylcaproic acid ≥97%
5-Methylcaproic acid

Hazard code(s): Irritant

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
Boiling Pt: 216 °C
MDL Number: MFCD00036494
CAS Number: 628-46-6
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Boiling point 216°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR