2,3,4,5-Tetrafluoroaniline ≥98%
Supplier: Matrix Scientific
|
Warning
|
003502-1G
003502-5G
101785-798EA
15.38
USD
101785-798
101785-800
2,3,4,5-Tetrafluoroaniline ≥98%
2,3,4,5-Tetrafluoroaniline
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₆H₃F₄N MW: 165.09 g/mol Boiling Pt: 78 °C (13mm) Melting Pt: 27…29 °C |
MDL Number:
MFCD00025153 CAS Number: 5580-80-3 |
Specification Test Results
Boiling point | 78°C/13mm |
Melting point | 27-29°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...