2,2,3,3-Tetrafluoro-1,4-butanediol ≥98%
Supplier: Matrix Scientific
Synonyms:
2,2,3,3-Tetrafluorobutane-1,4-diol
003476-5G
003476-25G
101785-754EA
89.23
USD
101785-754
101785-756
2,2,3,3-Tetrafluoro-1,4-butanediol ≥98%
2,2,3,3-Tetrafluoro-1,4-butanediol
Hazard code(s): Irritant
Formula:
C₄H₆F₄O₂ MW: 162.08 g/mol Boiling Pt: 110 - 112 °C (13 -rr) Melting Pt: 85 °C |
MDL Number:
MFCD00042375 CAS Number: 425-61-6 |
Specification Test Results
Boiling point | 110-112°C/1 |
Melting point | 85-87°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...