2,3,4,5-Tetrafluorobenzonitrile ≥97%

Supplier: Matrix Scientific

003454-1G 003454-5G 003454-25G
101785-726EA 23.08 USD
101785-726 101785-728 101785-730
2,3,4,5-Tetrafluorobenzonitrile ≥97%
2,3,4,5-Tetrafluorobenzonitrile

Hazard code(s): Irritant

Formula: C₇HF₄N
MW: 175.09 g/mol
Boiling Pt: 164 - 165 °C (1013 hPa)
Density: 1.463 g/cm³ (20 °C)
MDL Number: MFCD00040187
CAS Number: 16582-93-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR