2,3,5,6-Tetrachlorophenyl isothiocyanate for synthesis

Supplier: Matrix Scientific

003432-1G 003432-5G
101785-708EA 69.23 USD
101785-708 101785-710
2,3,5,6-Tetrachlorophenyl isothiocyanate for synthesis
2,3,5,6-Tetrachlorophenyl isothiocyanate

Hazard code(s): Lachrymator, Refrigerate

Formula: C₇HCl₄NS
Melting Pt: 88…90 °C
MDL Number: MFCD00060365
CAS Number: 22133-95-5

Order Now

Specification Test Results

Melting point 88-90°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR