4-Methyl-2,3,5,6-tetrabromophenol

Supplier: Matrix Scientific

Synonyms: 2,3,5,6-Tetrabromo-4-methylphenol, 2,3,5,6-Tetrabromo-p-cresol, 4-Hydroxy-2,3,5,6-tetrabromotoluene

003429-1G 003429-5G
101785-700EA 60 USD
101785-700 101785-702
4-Methyl-2,3,5,6-tetrabromophenol
4-Methyl-2,3,5,6-tetrabromophenol

Hazard code(s): Irritant-Harmful

Formula: C₇H₄Br₄O
MW: 423.72 g/mol
Boiling Pt: 356 °C (1013 hPa)
Melting Pt: 196 °C
MDL Number: MFCD00060318
CAS Number: 37721-75-8

Order Now

Specification Test Results

Melting point 196°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR