3'-Iodoacetophenone ≥99%
Supplier: Matrix Scientific
Synonyms:
1-(3-Iodophenyl)ethanone, m-Iodoacetophenone, 1-(3-Iodophenyl)ethan-1-one
003427-5G
003427-25G
003427-100G
101785-694EA
167.69
USD
101785-694
101785-696
101785-698
3'-Iodoacetophenone ≥99%
3'-Iodoacetophenone
Hazard code(s): Irritant
Formula:
C₈H₇IO MW: 246.05 g/mol Boiling Pt: 98 °C (1mm) Melting Pt: 53 °C Density: 1.767 g/cm³ (25 °C) |
MDL Number:
MFCD00060283 CAS Number: 14452-30-3 |
Specification Test Results
Boiling point | 98°C/1mm |
Density | 1.78 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...