4'-Iodoacetophenone ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone

003426-25G 003426-100G
101785-690EA 30.77 USD
101785-690 101785-692
4'-Iodoacetophenone ≥98% for synthesis
4'-Iodoacetophenone

Hazard code(s): Irritant

Formula: C₈H₇IO
Boiling Pt: 145…150 °C (20 mmHg)
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8

Order Now

Specification Test Results

Melting point 85-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR