o-Tolylacetic acid ≥99%
Supplier: Matrix Scientific
003420-25G
003420-100G
101785-678EA
18.46
USD
101785-678
101785-680
o-Tolylacetic acid ≥99%
o-Tolylacetic acid
Hazard code(s): Irritant
Formula:
(C₆H₄CH₃)CH₂COOH Boiling Pt: 283 °C (1013 hPa) Melting Pt: 88…90 °C |
MDL Number:
MFCD00004328 CAS Number: 644-36-0 EINECS: 211-416-9 |
Specification Test Results
Melting point | 88-90°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...