1-Chloro-4-ethylbenzene ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Chloro(ethylbenzene)

003406-5G 003406-25G
101785-666EA 106.15 USD
101785-666 101785-668
1-Chloro-4-ethylbenzene ≥97%
1-Chloro-4-ethylbenzene

Hazard code(s): Flammable

Formula: C₈H₉Cl
MW: 140.61 g/mol
Boiling Pt: 184…185 °C
MDL Number: MFCD00040855
CAS Number: 622-98-0
EINECS: 210-763-3

Order Now

Specification Test Results

Boiling point 184-185°C
ND20 1.5175

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR