2-Methoxyethyl isothiocyanate ≥98%

Supplier: Matrix Scientific

003383-5G 003383-25G
101785-646EA 52.31 USD
101785-646 101785-648
2-Methoxyethyl isothiocyanate ≥98%
2-Methoxyethyl isothiocyanate

Hazard code(s): Irritant, Lachrymator, Refrigerate

Formula: C₄H₇NOS
MW: 117.17 g/mol
Boiling Pt: 88.5 - 92 °C (23.5 -rr)
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00041126
CAS Number: 38663-85-3
EINECS: 254-070-4

Order Now

Specification Test Results

Boiling point 88-92°C/24m
Density 1.08
ND20 1.513

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR