3-Phenylbenzylamine ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-(Aminomethyl)biphenyl, [1,1'-Biphenyl]-3-ylmethanamine

003347-1G 003347-5G
101785-596EA 126.15 USD
101785-596 101785-598
3-Phenylbenzylamine ≥98% for synthesis
3-Phenylbenzylamine

Hazard code(s): Corrosive, Irritant

Formula: C₁₃H₁₃N
Boiling Pt: 128 °C (1mm)
Melting Pt: 28…30 °C
MDL Number: MFCD00270124
CAS Number: 177976-49-7

Order Now

Specification Test Results

Boiling point 128°C/1mm
Melting point 28-30°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR