4-Phenylbenzylamine ≥98% for synthesis

Supplier: Matrix Scientific

003345-1G 003345-5G
101785-592EA 69.23 USD
101785-592 101785-594
4-Phenylbenzylamine ≥98% for synthesis
4-Phenylbenzylamine

Hazard code(s): Irritant

Formula: C₁₃H₁₃N
Melting Pt: 55…57 °C
MDL Number: MFCD01310838
CAS Number: 712-76-5

Order Now

Specification Test Results

Melting point 55-57°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR