2-Fluorobenzyl isothiocyanate ≥98%

Supplier: Matrix Scientific

003337-1G 003337-5G
101785-588EA 29.23 USD
101785-588 101785-590
2-Fluorobenzyl isothiocyanate ≥98%
2-Fluorobenzyl isothiocyanate

Hazard code(s): Irritant-Harmful, Lachrymator, Refrigerate

Formula: C₈H₆FNS
MW: 167.21 g/mol
Boiling Pt: 130 °C (10mm)
Density: 1.22 g/cm³
MDL Number: MFCD00046817
CAS Number: 64382-80-5

Order Now

Specification Test Results

Boiling point 130°C/10mm
Density 1.22

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR