3-Chlorobenzyl alcohol ≥98%

Supplier: Matrix Scientific

003326-25G 003326-100G
101785-584EA 44.62 USD
101785-584 101785-586
3-Chlorobenzyl alcohol ≥98%
3-Chlorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₇ClO
Boiling Pt: 237 °C
Density: 1.211 g/cm³
MDL Number: MFCD00004632
CAS Number: 873-63-2
EINECS: 212-847-5

Order Now

Specification Test Results

Boiling point 237°C
Density 1.211
ND20 1.555


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR