2-Chlorobenzyl alcohol ≥99%

Supplier: Matrix Scientific

Synonyms: (2-Chlorophenyl)methanol

003325-25G 003325-100G
101785-580EA 20 USD
101785-580 101785-582
2-Chlorobenzyl alcohol ≥99%
2-Chlorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₇ClO
MW: 142.58 g/mol
Boiling Pt: 227 °C (1013 hPa)
Melting Pt: 68 - 71 °C
MDL Number: MFCD00004604
CAS Number: 17849-38-6
EINECS: 241-801-7

Order Now

Specification Test Results

Boiling point 227°C
Melting point 69-71°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR