4-Fluorobenzophenone ≥97%

Supplier: Matrix Scientific
Warning

003300-100G 003300-500G
101785-554EA 53.85 USD
101785-554 101785-556
4-Fluorobenzophenone ≥97%
4-Fluorobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₉FO
MW: 200.21 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 47…49 °C
MDL Number: MFCD00000352
CAS Number: 345-83-5
EINECS: 206-463-7

Order Now

Specification Test Results

Melting point 47-49°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR