3-Fluorobenzonitrile ≥98%

Supplier: Matrix Scientific

003297-25G 003297-100G
101785-544EA 20 USD
101785-544 101785-546
3-Fluorobenzonitrile ≥98%
3-Fluorobenzonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₄FN
Boiling Pt: 182…183 °C (753mm)
Melting Pt: –16 °C
Density: 1.1334 g/cm³ (25 °C)
MDL Number: MFCD00001797
CAS Number: 403-54-3
EINECS: 206-963-5
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 182-183°C/7
Melting point -16°C
Density 1.133
ND20 1.5043


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR