3-Fluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

003293-100G 003293-500G
101785-534EA 26.15 USD
101785-534 101785-536
3-Fluorobenzoic acid ≥98%
3-Fluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₅FO₂
MW: 140.11 g/mol
Boiling Pt: 251 °C (1013 hPa)
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00002489
CAS Number: 455-38-9
EINECS: 207-248-0

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR