3-Ethoxybenzhydrazide ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Ethoxybenzoic hydrazide
003282-1G
003282-5G
101785-502EA
27.69
USD
101785-502
101785-504
3-Ethoxybenzhydrazide ≥98%
3-Ethoxybenzhydrazide
Hazard code(s): Irritant
Formula:
C₉H₁₂N₂O₂ MW: 180.21 g/mol Melting Pt: 101…103 °C |
MDL Number:
MFCD00017063 CAS Number: 27830-16-6 |
Specification Test Results
Melting point | 101-103°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...