2-Ethoxybenzhydrazide
Supplier: Matrix Scientific
Synonyms:
2-Ethoxybenzohydrazide, 2-Ethoxybenzoic hydrazide, 2-Ethoxybenzoic acid hydrazide, 2-Ethoxybenzene-1-carbohydrazide
003281-5G
003281-25G
101785-498EA
103.08
USD
101785-498
101785-500
2-Ethoxybenzhydrazide
2-Ethoxybenzhydrazide
Hazard code(s): Irritant
Formula:
C₉H₁₂N₂O₂ MW: 180.21 g/mol Melting Pt: 62…64 °C |
MDL Number:
MFCD00017050 CAS Number: 21018-13-3 |
Specification Test Results
Melting point | 62-64°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...