2-Fluorobenzhydrazide ≥98%
Supplier: Matrix Scientific
003278-5G
003278-25G
101785-488EA
29.23
USD
101785-488
101785-490
2-Fluorobenzhydrazide ≥98%
2-Fluorobenzhydrazide
Hazard code(s): Irritant
Formula:
C₇H₇FN₂O Melting Pt: 72…73 °C |
MDL Number:
MFCD00025112 CAS Number: 446-24-2 |
Specification Test Results
Melting point | 72-73°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...