4-Fluorobenzenesulfonamide ≥97%

Supplier: Matrix Scientific

003269-25G 003269-100G
101785-480EA 38.46 USD
101785-480 101785-482
4-Fluorobenzenesulfonamide ≥97%
4-Fluorobenzenesulphonamide

Hazard code(s): Irritant

Formula: C₆H₆FNO₂S
MDL Number: MFCD00025384
CAS Number: 402-46-0
EINECS: 206-946-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR