4-Chlorobenzamidine hydroiodide ≥96%

Supplier: Matrix Scientific

Synonyms: 4-Chlorobenzene-1-carboximidamide hydroiodide, 4-Chlorophenylamidine hydroiodide

003264-1G 003264-5G
101785-472EA 44.62 USD
101785-472 101785-474
4-Chlorobenzamidine hydroiodide ≥96%
4-Chlorobenzamidine hydroiodide

Hazard code(s): Moisture Sensitive

Formula: C₇H₈ClIN₂
MW: 282.51 g/mol
Melting Pt: 237…240 °C
MDL Number: MFCD00051987
CAS Number: 115297-57-9

Order Now

Specification Test Results

Melting point 237-240°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR