2-Fluorobenzamide ≥98%

Supplier: Matrix Scientific

003261-25G 003261-100G
101785-466EA 38.46 USD
101785-466 101785-468
2-Fluorobenzamide ≥98%
2-Fluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₆FNO
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 115 - 119 °C
MDL Number: MFCD00007970
CAS Number: 445-28-3
EINECS: 207-157-6

Order Now

Specification Test Results

Melting point 117-119°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR