4-Bromophenylboronic acid ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Bromobenzeneboronic acid, (4-Bromophenyl)boronic acid, Dihydroxy(4-bromophenyl)borane, (4-Bromophenyl)metaboric acid

003243-5G 003243-25G
101785-440EA 38.46 USD
101785-440 101785-442
4-Bromophenylboronic acid ≥97%
4-Bromophenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Melting Pt: 282…286 °C
Density: 1.67 g/cm³ (20 °C)
MDL Number: MFCD00002104
CAS Number: 5467-74-3
EINECS: 226-779-9

Order Now

Specification Test Results

Melting point 282-286°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR