(3-Bromophenyl)acetic acid ≥98%
Supplier: Matrix Scientific
003236-25G
003236-100G
101785-428EA
73.85
USD
101785-428
101785-430
(3-Bromophenyl)acetic acid ≥98%
(3-Bromophenyl)acetic acid
Hazard code(s): Irritant
Formula:
(C₆H₄Br)CH₂COOH Boiling Pt: 308 °C (1013 hPa) Melting Pt: 100…102 °C |
MDL Number:
MFCD00004330 CAS Number: 1878-67-7 EINECS: 217-522-1 |
Specification Test Results
Melting point | 100-102°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...