4-Bromophenyl methyl sulfone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-(methylsulphonyl)benzene

003218-5G 003218-25G
101785-414EA 12.31 USD
101785-414 101785-416
4-Bromophenyl methyl sulfone ≥98%
4-Bromophenyl methyl sulphone

Hazard code(s): Irritant

Formula: C₇H₇BrO₂S
MW: 235.1 g/mol
Melting Pt: 101…102 °C
MDL Number: MFCD00025065
CAS Number: 3466-32-8
EINECS: 222-421-0

Order Now

Specification Test Results

Melting point 101-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR