2-(4-Aminophenyl)ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Aminophenethyl alcohol

003181-25G 003181-100G
101785-368EA 89.23 USD
101785-368 101785-370
2-(4-Aminophenyl)ethanol ≥95%
2-(4-Aminophenyl)ethanol

Hazard code(s): Irritant, Refrigerate, Stored under Argon

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 235…255 °C (1013 hPa)
Melting Pt: 108…110 °C
MDL Number: MFCD00007922
CAS Number: 104-10-9
EINECS: 203-174-8

Order Now

Specification Test Results

Melting point 108-110°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR