ω-Bromo-4-nitroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: α-Bromo-4-nitroacetophenone, 4-Nitrophenacyl bromide, Bromomethyl p-nitrophenyl ketone, 2-Bromo-4'-nitroacetophenone

003177-25G 003177-100G
101785-364EA 23.08 USD
101785-364 101785-366
ω-Bromo-4-nitroacetophenone ≥97%
ω-Bromo-4-nitroacetophenone

Hazard code(s): Corrosive, Lachrymator

Formula: C₈H₆BrNO₃
MW: 244.04 g/mol
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007356
CAS Number: 99-81-0
EINECS: 202-789-9

Order Now

Specification Test Results

Melting point 100-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR