1-Iodoperfluorododecane ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: Pentacosafluoro-1-iodododecane, Perfluorododecyl iodide

003153-1G 003153-5G
101785-322EA 110.77 USD
101785-322 101785-324
1-Iodoperfluorododecane ≥97%
1-Iodoperfluorododecane

Hazard code(s): Irritant

Formula: C₁₂F₂₅I
MW: 745.99 g/mol
Boiling Pt: 108…110 °C (18mm)
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00001066
CAS Number: 307-60-8
EINECS: 206-205-3

Order Now

Specification Test Results

Boiling point 108-110°C/1
Melting point 99-101°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR