2-Acetamido-5-fluorobenzoic acid

Supplier: Matrix Scientific

003146-1G 003146-5G
101785-306EA 43.08 USD
101785-306 101785-308
2-Acetamido-5-fluorobenzoic acid
2-Acetamido-5-fluorobenzoic acid
Formula: C₉H₈FNO₃
MDL Number: MFCD00973900
CAS Number: 49579-56-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR